Merck
  • Identification of Receptor Binding to the Biomolecular Corona of Nanoparticles.
Identification of Receptor Binding to the Biomolecular Corona of Nanoparticles.
ACS nano (2017-01-24)
Sandra Lara, Fatima Alnasser, Ester Polo, David Garry, Maria Cristina Lo Giudice, Delyan R Hristov, Louise Rocks, Anna Salvati, Yan Yan, Kenneth A Dawson
ABSTRACT
Biomolecules adsorbed on nanoparticles are known to confer a biological identity to nanoparticles, mediating the interactions with cells and biological barriers. However, how these molecules are presented on the particle surface in biological milieu remains unclear. The central aim of this study is to identify key protein recognition motifs and link them to specific cell-receptor interactions. Here, we employed an immuno-mapping technique to quantify epitope presentations of two major proteins in the serum corona, low-density lipoprotein and immunoglobulin G. Combining with a purpose-built receptor expression system, we show that both proteins present functional motifs to allow simultaneous recognition by low-density lipoprotein receptor and Fc-gamma receptor I of the corona. Our results suggest that the "labeling" of nanoparticles by biomolecular adsorption processes allows for multiple pathways in biological processes in which they may be "mistaken" for endogenous objects, such as lipoproteins, and exogenous ones, such as viral infections.
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
Gold(III) chloride trihydrate, ≥99.9% trace metals basis
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)