Merck
  • Modification of the existing maximum residue levels for etridiazole in various crops.
Modification of the existing maximum residue levels for etridiazole in various crops.
EFSA journal. European Food Safety Authority (2017-03-09)
Alba Brancato, Daniela Brocca, Chloe De Lentdecker, Zoltan Erdos, Lucien Ferreira, Luna Greco, Samira Jarrah, Dimitra Kardassi, Renata Leuschner, Christopher Lythgo, Paula Medina, Ileana Miron, Tunde Molnar, Alexandre Nougadere, Ragnor Pedersen, Hermine Reich, Angela Sacchi, Miguel Santos, Alois Stanek, Juergen Sturma, Jose Tarazona, Anne Theobald, Benedicte Vagenende, Alessia Verani, Laura Villamar-Bouza
ABSTRACT
In accordance with Article 6 of Regulation (EC) No 396/2005, the evaluating Member State (EMS), the Netherlands, received an application from Arysta LifeScience Great Britain Limited to modify the existing maximum residue levels (MRLs) for the active substance etridiazole in various crops. To accommodate for the intended uses of etridiazole, the Netherlands proposed to raise the existing MRLs from the limit of quantification (LOQ) of 0.05* mg/kg to 0.4 mg/kg in gherkin, courgette and other cucurbits with edible peel, and to raise the existing MRL value of 0.1 mg/kg to 0.4 mg/kg in cucumber. The Netherlands drafted an evaluation report in accordance with Article 8 of Regulation (EC) No 396/2005, which was submitted to the European Commission and forwarded to EFSA. Although the intended uses on gherkin, courgette and other cucurbits with edible peel are supported by residue trials, EFSA did not derive MRL proposals because appropriate information on the toxicological profile and on the relevance of the major plant metabolites 5-hydroxyethoxyetridiazole acid and 3-hydroxymethyletridiazole has not been provided. Thus, the tentative risk assessment is affected by a high degree of uncertainty.
Product Description
Supelco
Etridiazole, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.