Merck
  • Assessment of α-amylase and α-glucosidase inhibitory potential of Citrus reticulata peel extracts in hyperglycemic/hypoglycemic rats.
Assessment of α-amylase and α-glucosidase inhibitory potential of Citrus reticulata peel extracts in hyperglycemic/hypoglycemic rats.
3 Biotech (2021-04-06)
Sanaid Ghauri, Syed Qasim Raza, Muhammad Imran, Shagufta Saeed, Muhammad Rashid, Rahat Naseer
ABSTRACT
Diabetes mellitus is a metabolic disorder of carbohydrate metabolism. The management of Diabetes mellitus with phytochemicals is hallmark of this research. Citrus species are known for their health benefits and are used as traditional food in South East Asia. The total phenolic content of peels was analyzed using different solvents, while Gallic acid was used as standard. Both ethanolic, aqueous extracts of Citrus reticulata peel showed good inhibitory activity against amylase (90.67%, 15.33%) and moderate against glucosidase (70.8%, 14.8%), respectively. Sixteen rats were randomly divided into four groups (G1, G2, G3, and G4); G1 is a negative control (water), G4 is a positive control (Acarbose), while other two are experimental groups like G2 (fed with 100 mL and 20 mg/mL in hypoglycemic and hyperglycemic trials) and G3 fed with 200 mL and 40 mg/mL in hypoglycemic and hyperglycemic trials. A significant effect of treatments and value of time was found in hyperglycemic rats. Ethanolic extract showed a significant reduction in blood glucose levels in hypoglycemic (overnight fasting) rats which was comparable to the positive control. These results suggest that C. reticulata peels can contribute as a useful food ingredient as a potential antihyperglycemic agent in managing type 2 diabetes mellitus. In future, C. reticulata peel will be a good candidate for pharmaceutical industry.
Sigma-Aldrich
α-Glucosidase from Bacillus stearothermophilus, lyophilized powder, ≥50 units/mg protein
Pricing and availability is not currently available.