Merck
Collagen-induced arthritis in rats.
Methods in molecular medicine (2007-11-07)
Marie M Griffiths, Grant W Cannon, Tim Corsi, Van Reese, Kandie Kunzler
ABSTRACT
Collagen-Induced Arthritis (CIA) is a complex model of autoimmune-mediated arthritis that is regulated by multiple genetic and environmental factors. CIA is induced in rats by immunization with native type II collagen and develops joint pathology similar to that of rheumatoid arthritis. This chapter details methods for the extraction and purification of native type II collagen from sternal and articular cartilage, an arthritis induction protocol that has resulted in reproducible CIA expression in several rat strains from year to year and criteria for measuring clinical, radiographic and immunological outcome parameters characteristic of CIA.
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Anti-Rat IgG (whole molecule)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Pepstatin A, microbial, Purity ≥98%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%