Merck
  • AAV-glycine receptor α3 alleviates CFA-induced inflammatory pain by downregulating ERK phosphorylation and proinflammatory cytokine expression in SD rats.
AAV-glycine receptor α3 alleviates CFA-induced inflammatory pain by downregulating ERK phosphorylation and proinflammatory cytokine expression in SD rats.
Molecular medicine (Cambridge, Mass.) (2023-02-17)
Hung-Chen Wang, Kuang-I Cheng, Kuang-Yi Tseng, Aij-Lie Kwan, Lin-Li Chang
ABSTRACT
Glycine receptors (GlyRs) play key roles in the processing of inflammatory pain. The use of adeno-associated virus (AAV) vectors for gene therapy in human clinical trials has shown promise, as AAV generally causes a very mild immune response and long-term gene transfer, and there have been no reports of disease. Therefore, we used AAV for GlyRα1/3 gene transfer in F11 neuron cells and into Sprague-Dawley (SD) rats to investigate the effects and roles of AAV-GlyRα1/3 on cell cytotoxicity and inflammatory response. In vitro experiments were performed using plasmid adeno-associated virus (pAAV)-GlyRα1/3-transfected F11 neurons to investigate the effects of pAAV-GlyRα1/3 on cell cytotoxicity and the prostaglandin E2 (PGE2)-mediated inflammatory response. In vivo experiment, the association between GlyRα3 and inflammatory pain was analyzed in normal rats after AAV-GlyRα3 intrathecal injection and after complete Freund's adjuvant (CFA) intraplantar administration. Intrathecal AAV-GlyRα3 delivery into SD rats was evaluated in terms of its potential for alleviating CFA-induced inflammatory pain. The activation of mitogen-activated protein kinase (MAPK) inflammatory signaling and neuronal injury marker activating transcription factor 3 (ATF-3) were evaluated by western blotting and immunofluorescence; the level of cytokine expression was measured by ELISA. The results showed that pAAV/pAAV-GlyRα1/3 transfection into F11 cells did not significantly reduce cell viability or induce extracellular signal-regulated kinase (ERK) phosphorylation or ATF-3 activation. PGE2-induced ERK phosphorylation in F11 cells was repressed by the expression of pAAV-GlyRα3 and administration of an EP2 inhibitor, GlyRαs antagonist (strychnine), and a protein kinase C inhibitor. Additionally, intrathecal AAV-GlyRα3 administration to SD rats significantly decreased CFA-induced inflammatory pain and suppressed CFA-induced ERK phosphorylation, did not induce obvious histopathological injury but increased ATF-3 activation in dorsal root ganglion (DRGs). Antagonists of the prostaglandin EP2 receptor, PKC, and glycine receptor can inhibit PGE2-induced ERK phosphorylation. Intrathecal AAV-GlyRα3 administration to SD rats significantly decreased CFA-induced inflammatory pain and suppressed CFA-induced ERK phosphorylation, did not significantly induce gross histopathological injury but elicited ATF-3 activation. We suggest that PGE2-induced ERK phosphorylation can be modulated by GlyRα3, and AAV-GlyRα3 significantly downregulated CFA-induced cytokine activation.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Neurofilament 200 kDa Antibody, clone RT97, clone RT97, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®