Merck
  • Characterization of the human fetal gonad and reproductive tract by single-cell transcriptomics.
Characterization of the human fetal gonad and reproductive tract by single-cell transcriptomics.
Developmental cell (2024-02-01)
Jasin Taelman, Sylwia M Czukiewska, Ioannis Moustakas, Yolanda W Chang, Sanne Hillenius, Talia van der Helm, Lotte E van der Meeren, Hailiang Mei, Xueying Fan, Susana M Chuva de Sousa Lopes
ABSTRACT
During human fetal development, sex differentiation occurs not only in the gonads but also in the adjacent developing reproductive tract. However, while the cellular composition of male and female human fetal gonads is well described, that of the adjacent developing reproductive tract remains poorly characterized. Here, we performed single-cell transcriptomics on male and female human fetal gonads together with the adjacent developing reproductive tract from first and second trimesters, highlighting the morphological and molecular changes during sex differentiation. We validated different cell populations of the developing reproductive tract and gonads and compared the molecular signatures between the first and second trimesters, as well as between sexes, to identify conserved and sex-specific features. Together, our study provides insights into human fetal sex-specific gonadogenesis and development of the reproductive tract beyond the gonads.
Sigma-Aldrich
Anti-N-Cadherin antibody, Mouse monoclonal, clone GC-4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Collagen Type IV Antibody, Chemicon®, from rabbit