Merck
  • Myosin heavy chain isoform composition and Ca(2+) transients in fibres from enzymatically dissociated murine soleus and extensor digitorum longus muscles.
Myosin heavy chain isoform composition and Ca(2+) transients in fibres from enzymatically dissociated murine soleus and extensor digitorum longus muscles.
The Journal of physiology (2009-11-04)
Juan C Calderón, Pura Bolaños, Carlo Caputo
ABSTRACT
Electrically elicited Ca(2+) transients reported with the fast Ca(2+) dye MagFluo-4 AM and myosin heavy chain (MHC) electrophoretic patterns were obtained in intact, enzymatically dissociated fibres from adult mice extensor digitorum longus (EDL) and soleus muscles. Thirty nine fibres (23 from soleus and 16 from EDL) were analysed by both fluorescence microscopy and electrophoresis. These fibres were grouped as follows: group 1 included 13 type I and 4 type IC fibres; group 2 included 2 type IIC, 3 IIA and 1 I/IIA/IIX fibres; group 3 included 4 type IIX and 1 type IIX/IIB fibres; group 4 included 2 type IIB/IIX and 9 type IIB fibres. Ca(2+) transients obtained in groups 1, 2, 3 and 4 had the following kinetic parameters (mean +/- s.e.m.): amplitude (F/F): 0.61 +/- 0.05, 0.53 +/- 0.08, 0.61 +/- 0.06 and 0.61 +/- 0.03; rise time (ms): 1.64 +/- 0.05, 1.35 +/- 0.05, 1.18 +/- 0.06 and 1.14 +/- 0.04; half-amplitude width (ms): 19.12 +/- 1.85, 11.86 +/- 3.03, 4.62 +/- 0.31 and 4.23 +/- 0.37; and time constants of decay (tau(1) and tau(2), ms): 3.33 +/- 0.13 and 52.48 +/- 3.93, 2.69 +/- 0.22 and 41.06 +/- 9.13, 1.74 +/- 0.06 and 12.88 +/- 1.93, and 1.56 +/- 0.11 and 9.45 +/- 1.03, respectively. The statistical differences between the four groups and the analysis of the distribution of the parameters of Ca(2+) release and clearance show that there is a continuum from slow to fast, that parallels the MHC continuum from pure type I to pure IIB. However, type IIA fibres behave more like IIX and IIB fibres regarding Ca(2+) release but closer to type I fibres regarding Ca(2+) clearance. In conclusion, we show for the first time the diversity of Ca(2+) transients for the whole continuum of fibre types and correlate this functional diversity with the structural and biochemical diversity of the skeletal muscle fibres.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Triton X-100, BioXtra
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Polyacrylamide, nonionic water-soluble polymer
Sigma-Aldrich
Polyacrylamide, average Mn 150,000