Merck
  • Expression of the Pax2 transcription factor is associated with vestibular phenotype in the avian inner ear.
Expression of the Pax2 transcription factor is associated with vestibular phenotype in the avian inner ear.
Developmental neurobiology (2009-01-09)
Mark E Warchol, Guy P Richardson
ABSTRACT
The paired-domain transcription factor Pax2 is involved in many facets of inner ear development, but relatively little is known about the expression or function of Pax2 in the mature ear. In this study, we have used immunohistochemical methods to characterize the expression patterns of Pax2 in the sensory organs of inner ears from posthatch chicks. Immunoreactivity for Pax2 was observed in the nuclei of most hair cells and supporting cells in the vestibular organs. In contrast, Pax2 expression in the chick cochlea was limited to hair cells located in the very distal (low frequency) region. We then used organotypic cultures of the chick utricle to examine changes in Pax2 expression in response to ototoxic injury and during hair cell regeneration. Treatment with streptomycin resulted in the loss of most Pax2 immunoreactivity from the lumenal (hair cell) stratum of the utricle. During the early phases of regeneration, moderate Pax2 expression was maintained in the nuclei of proliferating supporting cells. Expression of Pax2 in the hair cell stratum recovered in parallel with hair cell regeneration. The results indicate that Pax2 continues to be expressed in the mature avian ear, and that its expression pattern is correlated with a vestibular phenotype.
Sigma-Aldrich
Phosphate buffered saline, pH 7.2 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, Autoclaved, pH 7.2 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, pH 7.4, contains BSA, powder
Sigma-Aldrich
Phosphate buffered saline, with 5% nonfat milk, powder (dry milled), pH 7.3
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
Phosphate buffered saline, BioUltra, pH 7.4 ( in solution), contains TWEEN 20, tablet
Sigma-Aldrich
Phosphate Buffered Saline, 10× PBS for Western blots and IP
Sigma-Aldrich
Phosphate Buffered Saline with 0.05% TWEEN® 20, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, BioPerformance Certified, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, BioUltra, solution
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 5 L solutions
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Phosphate Buffered Saline with 3% Non-Fat Milk, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, BioUltra, washing buffer for peroxidase conjugates in Western Blotting, 10x concentrate
Sigma-Aldrich
Phosphate buffered saline, pH 7.6 (25 °C)
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, BioUltra, ≥99%
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture