Merck
  • Indian muntjac, Muntiacus muntjak: a deer with a low diploid chromosome number.
Indian muntjac, Muntiacus muntjak: a deer with a low diploid chromosome number.
Science (New York, N.Y.) (1970-06-12)
D H Wurster, K Benirschke
ABSTRACT
The Indian muntjac (Muntiacus muntjak) has a diploid chromosome number of 7 in the male and 6 in the female, the lowest number yet described in a mammal. Its near relative, Reeve's muntjac (Muntiacus reevesi) has a diploid number of 46, and the karyotypes of the two, species are very different.