Merck
  • Studies on neurosteroids XXIII. Analysis of tetrahydrocorticosterone isomers in the brain of rats exposed to immobilization using LC-MS.
Studies on neurosteroids XXIII. Analysis of tetrahydrocorticosterone isomers in the brain of rats exposed to immobilization using LC-MS.
Steroids (2007-09-04)
Tatsuya Higashi, Hiroyuki Yokoi, Hitoe Maekubo, Ayako Honda, Kazutake Shimada
ABSTRACT
The identification and quantification of tetrahydrocorticosterone isomers (THBs; 3alpha,5alpha-, 3beta,5alpha-, 3alpha,5beta- and 3beta,5beta-THB) in rat brains using liquid chromatography (LC)-mass spectrometry (MS) are described. For the identification, the THBs were converted to the atmospheric pressure chemical ionization (APCI)-active derivatives, i.e., the dinitrobezoyl esters and 2-nitro-4-trifluoromethylphenyl hydrazones, and detected in the negative-ion mode. These derivatives showed 60- and 40-fold higher sensitivities, respectively, than intact steroids measured in the positive-APCI-MS. The derivatized THBs were satisfactorily separated from the others during the reversed-phase LC. The THBs were not detected at all in the brains of the unstressed rats. When the rats were exposed to the immobilization for 20 min, 3alpha,5alpha- and 3beta,5alpha-THB were detected as the major metabolites together with small amounts of 3alpha,5beta- and 3beta,5beta-THB in the male rat brain, while only 3alpha,5alpha-THB was detected in the female rats. Thus, the steroid variety found in the brains was different between the sexes. In the next step, 3alpha,5alpha-THB, a major metabolite found in the brains of the stressed rats, was quantified as its dinitrobezoyl ester. This method was accurate and reproducible, and the limit of quantitation was 1.0 ng/g tissue when a 50 mg tissue sample was used. There was also a sex difference in the brain 3alpha,5alpha-THB level; it was significantly higher in the female rats than in the male rats (P<0.05), although the brain corticosterone level was not higher in the stressed female rats than in the male rats (no statistical difference).