Merck
  • New prognosis biomarkers identified by dynamic proteomic analysis of colorectal cancer.
New prognosis biomarkers identified by dynamic proteomic analysis of colorectal cancer.
Molecular bioSystems (2012-09-22)
Ya Peng, Xiayu Li, Minghua Wu, Jing Yang, Minji Liu, Wengling Zhang, Bo Xiang, Xiaoyan Wang, Xiaoling Li, Guiyuan Li, Shourong Shen
ABSTRACT
The initiation, promotion and progression of human cancer are complex, polygenic, multi-factored processes. Through systematic proteomic analysis, different stages of CRC (colorectal cancer) biopsies were examined, and 199 differentially expressed proteins were detected between TNM (the tumor, nodes, and metastasis) stages I-IV and normal tissue (One-Way Analysis of Variance, ANOVA; p≤ 0.05). Instead of looking for biomarkers to distinguish CRC from normal or identify metastatic tumors, we focused on the variation tendency of CRC carcinogenesis and the dynamic expression patterns of proteins among the different stages. Som (self-organizing map clustering) analysis revealed eight unique expression patterns and that the cancer-related proteins were dynamically expressed, and their expression levels changed continuously throughout tumorigenesis. Molecular evidence emerged much earlier than visible, clinical or histological changes, which shows the potential prospect of building molecular staging. Proteins identified by MALDI-TOF MS (Matrix-Assisted Laser Desorption/Ionization Time of Flight Mass Spectrometry) were mainly involved in energy metabolism, acetylation and signaling pathways. Validation experiments using immunoblotting and immunohistochemistry (IHC) agreed with the 2D-DIGE (two-dimensional difference in gel electrophoresis) data. After survival classifier and LOOCV (leave-one-out cross-validation) analyses, the new prognostic biomarkers (78 kDa Glucose-Regulated Protein precursor (GRP78), Fructose-bisphosphate Aldolase A (ALDOA), Carbonic Anhydrase I (CA1) and Peptidyl-prolyl cis-trans isomerase A or Cyclophilin A (PPIA)) provided good survival prediction for TNM stage I-IV patients. The new biomarkers derived from the dynamic patterns of these proteins' expression provide is a good supplementary method for determining prognosis for CRC, especially for the TNM stage III and IV patients.
Sigma-Aldrich
Aldolase from rabbit muscle, ammonium sulfate suspension, 10-20 units/mg protein
Sigma-Aldrich
Aldolase from rabbit muscle, lyophilized powder, ≥8.0 units/mg protein