Merck
  • Resveratrol reduces matrix metalloproteinase-2 activity induced by oxygen-glucose deprivation and reoxygenation in human cerebral microvascular endothelial cells.
Resveratrol reduces matrix metalloproteinase-2 activity induced by oxygen-glucose deprivation and reoxygenation in human cerebral microvascular endothelial cells.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition (2013-04-18)
Zahide Cavdar, Mehtap Y Egrilmez, Zekiye S Altun, Nur Arslan, Nilgun Yener, Oya Sayin, Sermin Genc, Kursat Genc, Huray Islekel, Gulgun Oktay, Gul Guner Akdogan
ABSTRACT
The main pathophysiology in cerebral ischemia is the structural alteration in the neurovascular unit, coinciding with neurovascular matrix degradation. Among the human matrix metalloproteinases (MMPs), MMP-2 and -9, known as gelatinases, are the key enzymes for degrading type IV collagen, which is the major component of the basal membrane that surrounds the cerebral blood vessel. In the present study, we investigated the effects of resveratrol on cytotoxicity, reactive oxygen species (ROS), and gelatinases (MMP-2 and -9) in human cerebral microvascular endothelial cells exposed to 6 hours of oxygen-glucose deprivation and a subsequent 24 hours of reoxygenation with glucose (OGD/R), to mimic ischemia/reperfusion in vivo. Lactate dehydrogenase increased significantly, in comparison to that in the normoxia group. ROS was markedly increased in the OGD/R group, compared to normoxia. Correspondingly, ROS was significantly reduced with 50 μM of resveratrol. The proMMP-2 activity in the OGD/R group showed a statistically significant increase from the control cells. Resveratrol preconditioning decreased significantly the proMMP-2 in the cells exposed to OGD/R in comparison to that in the OGD/R group. Our results indicate that resveratrol regulates MMP-2 activity induced by OGD/R via its antioxidant effect, implying a possible mechanism related to the neuroprotective effect of resveratrol.
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, 1000 units/mL
Sigma-Aldrich
Lactic Dehydrogenase, recombinant from E. coli, ≥90 U/mg
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine muscle, Type X, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type XVII, buffered aqueous glycerol solution, ≥400 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type III, ammonium sulfate suspension, ≥500 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from porcine heart, ammonium sulfate suspension, ≥200 units/mg protein