Merck
  • Structural organization of the human neuronal nitric oxide synthase gene (NOS1).
Structural organization of the human neuronal nitric oxide synthase gene (NOS1).
The Journal of biological chemistry (1994-12-30)
A V Hall, H Antoniou, Y Wang, A H Cheung, A M Arbus, S L Olson, W C Lu, C L Kau, P A Marsden
ABSTRACT
Neuronal nitric oxide (NO) synthase, localized to human chromosome 12, uniquely participates in diverse biologic processes; neurotransmission, the regulation of body fluid homeostasis, neuroendocrine physiology, control of smooth muscle motility, sexual function, and myocyte/myoblast biology, among others. Restriction enzyme mapping, subcloning, and DNA sequence analysis of bacteriophage- and yeast artificial chromosome-derived human genomic DNA indicated that the mRNA for neuronal NO synthase is dispersed over a minimum of 160 kilobases of human genomic DNA. Analysis of intron-exon splice junctions predicted that the open reading frame is encoded by 28 exons, with translation initiation and termination in exon 2 and exon 29, respectively. Determination of transcription initiation sites in brain poly(A) RNA with primer extension analysis and RNase protection revealed a major start site 28 nucleotides downstream from a TATA box. Sequence inspection of 5'-flanking regions revealed potential cis-acting DNA elements: AP-2, TEF-1/MCBF, CREB/ATF/c-Fos, NRF-1, Ets, NF-1, and NF-kappa B-like sequences. Diversity appears to represent a major theme apparent upon analysis of human neuronal NO synthase mRNA transcripts. A microsatellite of the dinucleotide variety was detected within the 3'-untranslated region of exon 29. Multiple alleles were evident in normal individuals indicating the existence of allelic mRNA sequence variation. Characterization of variant human neuronal NO synthase cDNAs indicated the existence of casette exon 9/10 and exon 10 deletions as examples of structural mRNA diversity due to alternative splicing. The latter deletion of a 175-nucleotide exon introduces a frame-shift and premature stop codon indicating the potential existence of a novel NH2 terminus protein. In summary, analysis of the human neuronal NO synthase locus reveals a complex genomic organization and mRNA diversity that is both allelic and structural.
Sigma-Aldrich
Nitric Oxide Synthase, Inducible from mouse, recombinant, expressed in E. coli, buffered aqueous solution