Merck
  • Sonochemistry of quinones in argon-saturated aqueous solutions: enhanced cytochrome c reduction.
Sonochemistry of quinones in argon-saturated aqueous solutions: enhanced cytochrome c reduction.
Chemical research in toxicology (1999-09-22)
R C Lawson, A Ferrer, W Flores, A E Alegría
ABSTRACT
Sonolysis of argon-saturated aqueous quinone solutions resulted in an enhancement in ferricytochrome c (Cyt c) reduction. Upon addition of superoxide dismutase, Cyt c reduction was partially inhibited, thus implying a role of superoxide ion in this reduction process. Neither quinone hydrophobicity nor reduction potential exclusively controls the Cyt c reduction enhancement, although a preference for hydrophobicity versus reduction potential is noted.
Sigma-Aldrich
2,6-Dimethylbenzoquinone, 99%
Sigma-Aldrich
2,3-Dimethoxy-5-methyl-p-benzoquinone, apoptosis inducer