Merck
Biodegradation of p-toluenesulphonamide by a Pseudomonas sp.
FEMS microbiology letters (2001-12-04)
A M van Haperen, J W van Velde, C G van Ginkel
ABSTRACT
A bacterium capable of utilising p-toluenesulphonamide was isolated from activated sludge. The isolated strain designated PTSA was identified as a Pseudomonas sp. using chemotaxonomic and genetic studies. Pseudomonas PTSA grew on p-toluenesulphonamide in a chemostat with approximately 90% release of sulphate and 80% release of ammonium. The isolate was also able to grow on 4-carboxybenzenesulphonamide and 3,4-dihydroxybenzoate but did not grow on p-toluenesulphonate. The transient appearance of 4-hydroxymethylbenzenesulphonamide and 4-carboxybenzenesulphonamide during p-toluenesulphonamide degradation proves oxidation of the methyl group is the initial attack in the biodegradation pathway. Both metabolites of p-toluenesulphonamide degradation were identified by high-performance liquid chromatography-mass spectrometry. 4-Carboxybenzenesulphonamide is probably converted into 3,4-dihydroxybenzoate and amidosulphurous acid. The latter is a chemically unstable compound in aqueous solutions and immediately converted into sulphite and ammonium. Both sulphite and ammonium were formed during degradation of 4-carboxybenzenesulphonamide.
Sigma-Aldrich
p-Toluenesulfonamide, ReagentPlus®, ≥99%
Sigma-Aldrich
p-Toluenesulfonamide, reagent grade, 97%