Merck
  • Effects of various insecticides on the development of the egg parasitoid Trichogramma dendrolimi (Hymenoptera: Trichogrammatidae).
Effects of various insecticides on the development of the egg parasitoid Trichogramma dendrolimi (Hymenoptera: Trichogrammatidae).
Journal of economic entomology (2002-01-05)
Y Takada, S Kawamura, T Tanaka
ABSTRACT
The toxicity of six insecticides, acephate, methomyl, ethofenprox, cartap, chlorfluazuron, and Bacillus thuringiensis (Bt) was tested on different developmental stages of the egg parasitoid, Trichogramma dendrolimi (Matsumura). Each of the insecticides tested showed different degrees of toxicity to the parasitoid, Ethofenprox showed the highest toxicity and cartap showed relatively higher toxicity compared with the other insecticides. The development of the parasitoids treated with these two insecticides was normal, similar to that of the control group. Only the emergence of adult wasps from host eggs was disturbed. Emergence of the host, Mamestra brassicae larva was reduced following treatment with ethofenprox, cartap and methomyl. However, adult female wasps, which emerged from host eggs treated with the insecticides had the ability to oviposit normally.
Product Description
Supelco
Cartap hydrochloride, PESTANAL®, analytical standard
Supelco
Chlorfluazuron, PESTANAL®, analytical standard
Supelco
Etofenprox, PESTANAL®, analytical standard