Merck
  • 1,2-bis(1H-indol-3-yl)ethane-1,2-dione, an indole alkaloid from the marine sponge Smenospongia sp.
1,2-bis(1H-indol-3-yl)ethane-1,2-dione, an indole alkaloid from the marine sponge Smenospongia sp.
Journal of natural products (2002-04-27)
Matthew J McKay, Anthony R Carroll, Ronald J Quinn, John N A Hooper
ABSTRACT
1,2-Bis(1H-indol-3-yl)ethane-1,2-dione (1), a bisindole alkaloid, was isolated from the marine sponge Smenospongia sp. The known compounds indole-3-carbaldehyde (2), 6-bromoindole-3-carbaldehyde (3), and tryptamine (4) and mixtures of the (Z/E) isomers of 6-bromo-2'-demethylaplysinopsin (5a/5b) and 6-bromo-3'-deimino-2',4'-bis(demethyl)-3'-oxoaplysinopsin (6a/6b) were also isolated.
Sigma-Aldrich
Indole-3-carboxaldehyde, 97%