Merck
  • Effects of intrinsic fluorescence and quenching on fluorescence-based screening of natural products.
Effects of intrinsic fluorescence and quenching on fluorescence-based screening of natural products.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2002-06-06)
L Zou, M R Harkey, G L Henderson
ABSTRACT
To evaluate the effects of intrinsic (natural) fluorescence and quenching as confounding variables in fluorescence-based enzyme inhibition assays of natural products, we measured the fluorescence and quenching properties of 25 components of popular herbal products. The analyses were performed under conditions typically employed in drug-drug interaction studies that use c-DNA-derived P450 isoforms and surrogate fluorogenic substrates. Four of the 25 compounds tested (isorhamnetin, quercetin, vitexin, and yangonin) fluoresced or quenched sufficiently to interfere with these assays. Intrinsic fluorescence had a greater effect on these assays than quenching and for one compound, yangonin, was sufficient to mask inhibition and potentially produce a false negative result. Quenching had less of an effect on these assays, but was significant enough for one compound, quercetin, to mimic "weak" inhibition. Therefore, because intrinsic fluorescence or quenching could render some natural products unsuitable for testing in certain fluorometric assays, it would be prudent to include an evaluation of these properties in experimental protocols.