Merck
  • Determination of delta9-THC in whole blood using gas chromatography-mass spectrometry.
Determination of delta9-THC in whole blood using gas chromatography-mass spectrometry.
Journal of analytical toxicology (2002-12-28)
Mark Hok Chi Chu, Olaf H Drummer
ABSTRACT
A simple and reliable liquid-liquid extraction method for the determination of delta9-tetrahydrocannabinol (THC) in whole blood utilizing gas chromatography-mass spectrometry in electron impact mode is described. The substance is derivatized with pentafluoropropionic anhydride in pentafluropropanol. The limit of detection is 0.5 ng/mL for a 1-mL specimen, with recovery greater than 70%. The intra-assay coefficient of variation (CV) is 3.1% to 5.2%, and the interassay CV is 6.4% to 9.5%, calculated at THC concentrations of 1, 5, and 25 ng/mL. The accuracy is between 95 and 97%. The optimization of extraction and derivatization conditions is detailed.
Sigma-Aldrich
Pentafluoropropionic anhydride, purum, ≥97.0% (GC)
Sigma-Aldrich
Pentafluoropropionic anhydride, 99%
Supelco
Pentafluoropropionic anhydride, derivatization grade (GC derivatization), LiChropur, 99%