Merck
A rapid colorimetric assay for gamma-glutamyl hydrolase (conjugase).
Analytical biochemistry (1992-11-15)
P V Wagh, T I Kalman
ABSTRACT
A simple procedure for the measurement of gamma-glutamyl hydrolase (conjugase) activity is described. Glutamic acid released from pteroylpenta-gamma-glutamate by hog kidney and chicken pancreas conjugases was quantitated using the dye 4,4'-bis(dimethylamino)benzophenone hydrazone. The procedure involves hydrolysis of the folylpoly-gamma-glutamate substrate by conjugase, conversion of glutamate to alpha-ketoglutarate by L-glutamate dehydrogenase and colorimetric measurement of the BDBH derivative of alpha-ketoglutarate. The release of as little as one nmol of glutamic acid from the substrate can be measured by this procedure, which is well suited for the assay of a variety of conjugase preparations. In addition, the method should provide a general assay for the enzymatic hydrolysis of various folate and antifolate polyglutamates.
Sigma-Aldrich
Michler′s ketone, 98%