Merck
  • Reaction products of W(CO)(6) with formamidines; electronic structure of a W(2)(mu-CO)(2) core with unsymmetric bridging carbonyls.
Reaction products of W(CO)(6) with formamidines; electronic structure of a W(2)(mu-CO)(2) core with unsymmetric bridging carbonyls.
Inorganic chemistry (2004-10-27)
F Albert Cotton, James P Donahue, Michael B Hall, Carlos A Murillo, Dino Villagrán
ABSTRACT
Reactions of W(CO)(6) with formamidines contrast with those of Mo(CO)(6) and Cr(CO)(6) in that the former do not yield quadruply bonded dimetal species. From the reaction of W(CO)(6) with HDAniF (HDAniF = N,N'-di-p-anisylformamidine), several new ditungsten carbonyl compounds (W(2)(mu-CO)(2)(mu-DAniF)(2)(eta(2)-DAniF)(2) (1), W(2)(mu-CO)(2)(mu-DAniF)(2)(eta(2)-DAniF)(eta(2)-CH(2)DAniF) (2), and W(2)(mu-CO)(mu-CNC(6)H(4)OCH(3))(mu-DAniF)(2)(eta(2)-DAniF)(2) (3)) have been isolated and fully characterized. In 2, CH(2)DAniF represents a DAniF ligand in which a methylene group has been added to one of the nitrogen atoms. This ligand binds to the tungsten atom using a nitrogen and a carbon atom. Compound 1 has a tungsten-tungsten bond distance of 2.476(1) A and a planar W(2)(mu-CO)(2) core structure which has C(2)(h)() symmetry with short and long W-C bond distances (1.99(1) and 2.28(1) A, respectively). DFT calculations on a model of 1 indicate that (a) the C(2)(h)() instead of D(2)(h)() symmetry of the ditungsten core may be attributed to W --> CO pi back-bonding interactions and (b) the bond between the tungsten atoms may be formulated as a double bond. The new tetragonal paddlewheel compound W(2)(DAniF)(4) (4) and the edge-sharing bioctahedron W(2)(mu-O)(mu-NC(6)H(3)Cl(2))(mu-D(Cl)PhF)(2)(eta(2)-D(Cl)PhF)(2) (5) (D(Cl)PhF = N,N'-di-(3,5-dichlorophenyl)formamidinate) have also been prepared.
Sigma-Aldrich
Formamidine hydrochloride, 97%
Sigma-Aldrich
Formamidine acetate salt, 99%