Merck
  • Brain lipid peroxidation and antioxidant status after acute methacrylonitrile intoxication.
Brain lipid peroxidation and antioxidant status after acute methacrylonitrile intoxication.
Drug and chemical toxicology (2005-05-04)
A Zeenath Unnisa, S Niranjali Devaraj
ABSTRACT
The status of brain antioxidant enzymes and glutathione in methacrylonitrile (MeAN)-intoxicated Wistar rats was correlated with the levels of lipid peroxidation products. Optimum changes were observed 30 min and 60 min after oral administration of MeAN at dosages of 50 mg/kg body weight per day (0.25 LD50) and 100 mg/kg body weight per day (0.5 LD50). An increase in lipid peroxidation products, decrease in the activity of antioxidant enzymes such as superoxide dismutase (SOD), catalase (CAT), glutathione peroxidase (GPX), glutathione reductase (GR), glutathione S-transferase (GST), and decrease in reduced glutathione (GSH) were observed. These studies suggest that the membrane lipid peroxidation observed in MeAN intoxication is related, in part, to a compromised antioxidant defense system.