Merck
  • Diagnosis of induced cleft palate in rats from defective patterns of differentiation of isoenzymes.
Diagnosis of induced cleft palate in rats from defective patterns of differentiation of isoenzymes.
Scandinavian journal of plastic and reconstructive surgery and hand surgery (1992-01-01)
G Granström, C Jacobsson, S Kirkeby
ABSTRACT
Cleft palate was induced in 420 embryos of Sprague-Dawley rats with a single oral dose of 600 mg/kg beta-aminoproprionitrile (BAPN) on embryonal day 15, 7 hours. The cleft palate was accompanied by a pathological differentiation pattern of various isoenzymes in palatal shelves. These isoenzymes could be detected in amniotic fluid from the 16th to the 20th days of pregnancy when they also had a pathological differentiation pattern. We conclude that teratogenically induced cleft palate in rats is accompanied by a pathological differentiation pattern that can be traced by determination of isoenzymes in the palatal shelves as well as in amniotic fluid.
Sigma-Aldrich
3-Aminopropionitrile fumarate salt, metabolite