Merck
  • [GC-MS analysis of the fatty components of pollen Typhae before and after being carbonized].
[GC-MS analysis of the fatty components of pollen Typhae before and after being carbonized].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2006-04-01)
Hong-Fei Ma, Bin Liu, Gui-Yan Zhang, Ren-Bing Shi, Chang-Hua Ma, Mi-Mi Yu
ABSTRACT
To study the changes of the fatty components of Pollen Typhae before and after being carbonized. Pollen Typhae and Pollen Typhae carbonisatus were extracted with petroleum ether (60-90 degrees C) respectively. The two kinds of extracts were analyzed by GC-MS after saponificated and methanolized, and their constituents were searched through NIST. The contents of the constituents were determined by method of normalization. Either in Pollen Typhae or in Pollen Typhae carbonisatus, 32 components were identified, among which 20 components were the same and 6 were different respectively. Among the same components, the relative contents of 3-methyl-2-butenoic acid-2-phenylethyl ester, hexanedioic acid-dimethyl ester, dimethyl phthalate, diethyl phthalate, diphenylamine, sebacic acid dimethyl ester, 1,2-benzenedicarboxylic acid, ethyl methyl ester, methyl-2-ethylhexyl phthalate and diisooctyl phthalate etc. increased obviously, and the relative contents of nonanedioic acid-dimethyl ester, diisobutyl phthalate and stigmastan-3,5-dien etc. decreased greatly. Among the different components, 8-hydroxy-octanoic acid-methyl ester, 9-hydroxy-nonanoic acid-methyl ester, 10-octadecenoic acid-methyl ester, m-hydroxycinnamic acid-methyl ester,3-[4-( acetyloxy)-3-methoxyphenyl]2-propenoic acid-methyl ester and 11-octadecenoic acid-methyl ester were detected in Pollen Typhae, 3-hydroxyspirost-8-en-11-one, benzenepropanoic acid-methyl ester, 2,4-dimethylhexanedioic acid; 2,4-bis (1,1-dimethylethyl)-phenol, undecanedioic acid-dimethyl ester and 9,10-dihydroxy-octadecanoic acid-methyl ester were detected in Pollen Typhae carbonistatus. The species and contents of the fatty components in Pollen Typhae changed before and after being carbonized, but their chemical types didn't change too much.
Product Description
Supelco
Diisobutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Diisobutyl phthalate, 99%