- Investigation into the temporal stability of aqueous standard solutions of psilocin and psilocybin using high performance liquid chromatography.
Investigation into the temporal stability of aqueous standard solutions of psilocin and psilocybin using high performance liquid chromatography.
Science & justice : journal of the Forensic Science Society (2006-09-28)
N Anastos, N W Barnett, F M Pfeffer, S W Lewis
PMID17002211
ABSTRACT
This paper reports an investigation into the temporal stability of aqueous solutions of psilocin and psilocybin reference drug standards over a period of fourteen days. This study was performed using high performance liquid chromatography utilising a (95:5% v/v) methanol: 10 mM ammonium formate, pH 3.5 mobile phase and absorption detection at 269 nm. It was found that the exclusion of light significantly prolonged the useful life of standards, with aqueous solutions of both psilocin and psilocybin being stable over a period of seven days.