Merck
  • Ferric iron is genotoxic in non-transformed and preneoplastic human colon cells.
Ferric iron is genotoxic in non-transformed and preneoplastic human colon cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2006-12-13)
Y Knöbel, A Weise, M Glei, W Sendt, U Claussen, B L Pool-Zobel
ABSTRACT
Iron could be a relevant risk factor for carcinogenesis since it catalyses the formation of reactive oxygen species (ROS), which damage DNA. We previously demonstrated genotoxic effects by ferric iron using the human colon cancer cell line HT29. Here we investigated ferric iron in primary non-transformed colon cells and in a preneoplastic colon adenoma cell line (LT97), which both are suitable models to study effects of carcinogens during early stages of cell transformation. Genetic damage was determined using the Comet assay. Comet FISH (fluorescence in situ hybridization) was used to assess specific effects on TP53. Fe-NTA (0-1000 microM, 30 min, 37 degrees C) significantly induced single strand breaks in primary colon cells (500 microM Fe-NTA: Tail intensity [TI] 22.6%+/-5.0% versus RPMI control: TI 10.6%+/-3.9%, p<0.01) and in LT97 cells (1000 microM Fe-NTA: TI 26.8%+/-7.3% versus RPMI control: TI 11.1%+/-3.7%, p<0.01). With the Comet FISH protocol lower concentrations of Fe-NTA significantly increased DNA damage already at 100 and 250 microM Fe-NTA in primary colon and LT97 adenoma cells, respectively. This damage was detected as an enhanced migration of TP53 signals into the comet tail in both cell types, which indicates a high susceptibility of this tumor relevant gene towards Fe-NTA. In conclusion, Fe-NTA acts genotoxic in non-transformed and in preneoplastic human colon cells, in which it also enhances migration of TP53 at relatively low concentrations. Translated to the in vivo situation these results suggest that iron overload putatively contributes to a genotoxic risk during early stages of colorectal carcinogenesis on account of its genotoxic potential in non-tumorigenic human colon cells.
Sigma-Aldrich
Iron(III) nitrate nonahydrate, ≥99.999% trace metals basis
Sigma-Aldrich
Iron(III) nitrate nonahydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Iron(III) nitrate nonahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Iron(III) nitrate nonahydrate, BioReagent, suitable for cell culture