Merck
  • Hydrogen bonding-induced aromatic oligoamide foldamers as spherand analogues to accelerate the hydrolysis of nitro-substituted anisole in aqueous media.
Hydrogen bonding-induced aromatic oligoamide foldamers as spherand analogues to accelerate the hydrolysis of nitro-substituted anisole in aqueous media.
The Journal of organic chemistry (2007-01-27)
Hui-Ping Yi, Jiang Wu, Kui-Ling Ding, Xi-Kui Jiang, Zhan-Ting Li
ABSTRACT
Four intramolecular hydrogen bonding-driven aromatic amide foldamers 2-5 have been designed and synthesized in which a 2-methoxy-3-nitrobenzamide unit was incorporated at the end of the backbone. Kinetic studies in dioxane-water (4:1, v/v) at 60-90 degrees C have revealed that the folded backbone of the oligomers was, like the rigidified spherand, able to complex Li+, Na+, and K+ and, consequently, accelerated the hydrolysis of the nitro-appended anisole unit of the foldamers. Generally, longer foldamers displayed an increased accelerating effect, and LiOH displayed the highest reactivity probably due to the most efficient complexation by the folded oligomers. Addition of excessive potassium chloride substantially reduced the complexing interaction, and the hydrolysis of the longer oligomers became slower than that of the shorter ones due to an increased steric effect. The results indicate that, even in a hot aqueous medium of high polarity, intramolecular hydrogen bonding is still able to induce structurally matched oligomers to generate a preorganized rigidified conformation.
Sigma-Aldrich
Lithium hydroxide, powder, reagent grade, ≥98%
Sigma-Aldrich
Lithium hydroxide, reagent grade, 98%
Sigma-Aldrich
Lithium hydroxide monohydrate, 99.95% trace metals basis
Sigma-Aldrich
Lithium hydroxide monohydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Lithium hydroxide monohydrate, BioXtra, 98.5-101.5% (titration)
Sigma-Aldrich
Lithium hydroxide monohydrate, ACS reagent, ≥98.0%