Merck
  • Extractive colorimetric method for the determination of dothiepin hydrochloride and risperidone in pure and in dosage forms.
Extractive colorimetric method for the determination of dothiepin hydrochloride and risperidone in pure and in dosage forms.
Chemical & pharmaceutical bulletin (2008-08-02)
Wafaa El-Sayed Hassan
ABSTRACT
Three rapid, simple, reproducible and sensitive extractive colorimetric methods (A--C) for assaying dothiepin hydrochloride (I) and risperidone (II) in bulk sample and in dosage forms were investigated. Methods A and B are based on the formation of an ion pair complexes with methyl orange (A) and orange G (B), whereas method C depends on ternary complex formation between cobalt thiocyanate and the studied drug I or II. The optimum reaction conditions were investigated and it was observed the calibration curves resulting from the measurements of absorbance concentration relations of the extracted complexes were linear over the concentration range 0.1--12 microg ml(-1) for method A, 0.5--11 mug ml(-1) for method B, and 3.2--80 microg ml(-1) for method C with a relative standard deviation (RSD) of 1.17 and 1.28 for drug I and II, respectively. The molar absorptivity, Sandell sensitivity, Ringbom optimum concentration ranges, and detection and quantification limits for all complexes were calculated and evaluated at maximum wavelengths of 423, 498, and 625 nm, using methods A, B, and C, respectively. The interference from excipients commonly present in dosage forms and common degradation products was studied. The proposed methods are highly specific for the determination of drugs I and II, in their dosage forms applying the standard additions technique without any interference from common excipients. The proposed methods have been compared statistically to the reference methods and found to be simple, accurate (t-test) and reproducible (F-value).
Sigma-Aldrich
Orange G, certified by the BSC, Dye content 80 %
Sigma-Aldrich
Orange G, for NA electrophoresis
Sigma-Aldrich
Orange G, certified by the BSC
Supelco
Risperidone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cobalt(II) thiocyanate, 99.9% trace metals basis
Sigma-Aldrich
Cobalt(II) thiocyanate, 96%