Merck
  • Theoretical study of the nuO-H IR spectra for the hydrogen bond dimers from the polarized spectra of glutaric and 1-naphthoic acid crystals: Fermi resonances effects.
Theoretical study of the nuO-H IR spectra for the hydrogen bond dimers from the polarized spectra of glutaric and 1-naphthoic acid crystals: Fermi resonances effects.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2009-06-16)
Mohamed El Amine Benmalti, Abdelghani Krallafa, Najeh Rekik, Mostefa Belhakem
ABSTRACT
A full quantum theoretical model is proposed to study the nu(O-H) experimental IR line shapes of polarized crystalline glutaric and 1-naphthoic acid dimer crystals at room and liquid nitrogen temperatures. This work is an application of a previous model [M. E-A. Benmalti, D. Chamma, P. Blaise, and O. Henri-Rousseau, J. Mol. Struct. 785 (2006) 27-31] by accounting for Fermi resonances. The approach is dealing with the strong anharmonic coupling, Davydov coupling, multiple Fermi resonances between the first harmonics of some bending modes and the first excited state of the symmetric combination of the two nu(O-H) modes and the quantum direct and indirect relaxation. Numerical results show that mixing of all these effects allows to reproduce satisfactorily the main features of the experimental IR line shapes of crystalline hydrogenated and deuterated glutaric and 1-naphthoic acid crystals and are expected to provide efficient of Fermi resonances effects.
Sigma-Aldrich
1-Naphthoic acid, 96%
Sigma-Aldrich
Glutaric acid, 99%