Merck
Reliable synthesis of various nucleoside diphosphate glycopyranoses.
Chemistry (Weinheim an der Bergstrasse, Germany) (2009-07-02)
Saskia Wolf, Tanja Zismann, Nathalie Lunau, Chris Meier
ABSTRACT
A reliable and high yielding synthetic pathway for the synthesis of the biologically highly important class of nucleoside diphosphate sugars (NDP-sugars) was developed by using various cycloSal-nucleotides 1 and 9 as active ester building blocks. The reaction with anomerically pure pyranosyl-1-phosphates 2 led to the target NDP-sugars 20-45 in a nucleophilic displacement reaction, which cleaves the cycloSal moiety in anomerically pure forms. As nucleosides cytidine, uridine, thymidine, adenosine, 2'-deoxy-guanosine and 2',3'-dideoxy-2',3'-didehydrothymidine were used while the phosphates of D-glucose, D-galactose, D-mannose, D-NAc-glucosamine, D-NAc-galactosamine, D-fucose, L-fucose as well as 6-deoxy-D-gulose were introduced.