Merck
  • Cesium carbonate-catalyzed alpha-phenylchalcogenation of carbonyl compounds with diphenyl dichalcogenide.
Cesium carbonate-catalyzed alpha-phenylchalcogenation of carbonyl compounds with diphenyl dichalcogenide.
Molecules (Basel, Switzerland) (2009-09-29)
Yutaka Nishiyama, Yuya Koguma, Toshimasa Tanaka, Rui Umeda
ABSTRACT
It was found that cesium carbonate has a unique catalytic ability on the reaction of carbonyl compounds with diphenyl diselenide to give the corresponding alpha-phenylseleno carbonyl compounds in moderate to good yields. Similarly, the alpha-phenylthiolation of carbonyl compounds with diphenyl disulfide was promoted by the cesium carbonate catalyst.
Sigma-Aldrich
Phenyl disulfide, 99%
Sigma-Aldrich
Cesium carbonate, ReagentPlus®, 99%