Merck
  • Terahertz absorption spectroscopy of a liquid using a polarity probe: a case study of trehalose/water mixtures.
Terahertz absorption spectroscopy of a liquid using a polarity probe: a case study of trehalose/water mixtures.
Angewandte Chemie (International ed. in English) (2009-12-08)
Mohsen Sajadi, Yathrib Ajaj, Ilya Ioffe, Hermann Weingärtner, Nikolaus P Ernsting
Sigma-Aldrich
Quinaldine, ≥95.0% (GC)
Supelco
Quinaldine, analytical standard, ≥90% (GC)