Merck
  • Disaggregation of high-molecular weight species during downstream processing to recover functional monomer.
Disaggregation of high-molecular weight species during downstream processing to recover functional monomer.
Biotechnology progress (2009-12-30)
Xuankuo Xu, David M Didio, Kirk J Leister, Sanchayita Ghose
ABSTRACT
The use of chaotropic agents to recover functional monomeric material was investigated for the downstream purification of an Fc-fusion protein containing high levels of high-molecular weight (HMW) species. In batch studies, chaotropic agents irreversibly disaggregated a majority of the aggregated protein. An integrated processing mode, termed as on-column disaggregation, was developed in which the protein was captured on Protein A chromatography and then a chaotropic agent was used to simultaneously elute the bound protein and disaggregate the HMW species. On-column disaggregation process resulted in protein recoveries of >95% and aggregation reduction of approximately 50%. Analytical results are presented showing that the recovered monomeric material was comparable to the reference protein in biochemical, biophysical, and pharmacokinetic properties. The kinetic and molecular mechanisms governing protein aggregation and disaggregation will also be elucidated. For the Fc-fusion protein studied here, incorporation of the disaggregation strategy in both batch and on-column modes led to an increase of >10% in overall downstream yield.
Sigma-Aldrich
Sodium thiocyanate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium thiocyanate solution, BioUltra, 8 M in H2O
Sigma-Aldrich
Sodium thiocyanate, ACS reagent, ≥98.0%
Sigma-Aldrich
Sodium thiocyanate, reagent grade, 98-102% (titration)