Merck
  • Application of ex situ dynamic nuclear polarization in studying small molecules.
Application of ex situ dynamic nuclear polarization in studying small molecules.
Physical chemistry chemical physics : PCCP (2010-05-11)
Christian Ludwig, Ildefonso Marin-Montesinos, Martin G Saunders, Abdul-Hamid Emwas, Zoe Pikramenou, Stephen P Hammond, Ulrich L Günther
ABSTRACT
Dynamic nuclear polarization (DNP) has become an attractive technique to boost the sensitivity of NMR experiments. In the case of ex situ polarizations two-dimensional (2D) spectra are limited by the short lifetime of the polarization after dissolution and sample transfer to a high field NMR magnet. This limitation can be overcome by various approaches. Here we show how the use of (13)C-labelled acetyl tags can help to obtain 2D-HMQC spectra for many small molecules, owing to a nuclear Overhauser enhancement between (13)C spins originating from the long-lived carbonyl carbon, which extends the lifetimes of other (13)C spins with shorter longitudinal relaxation times. We also show an alternative approach of using an optimized polarization matrix.
Sigma-Aldrich
4-Methoxyphenol, purum, ≥98.0% (HPLC)
Sigma-Aldrich
4-Methoxyphenol, ReagentPlus®, 99%
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%
Sigma-Aldrich
Acetyl chloride, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%