Merck
  • Transesterification/acetylation of long chain alcohols with alkyl acetate.
Transesterification/acetylation of long chain alcohols with alkyl acetate.
Chemistry and physics of lipids (2010-07-06)
Phattaraporn Kaewkool, Kanit Krisnangkura
ABSTRACT
Gas chromatographic characterizations of fatty alcohols are generally carried out as the free alcohols, trimethyl silyl or acetyl derivatives. In this study, transesterification/acetylation of long chain fatty alcohols is simply carried out by dissolving the alcohol in ethyl/methyl acetate and passing through a micro-column packed with solid NaOH. Reaction times are slightly different for alcohols of different chain length. Rice bran alcohols of 24-34 carbon atom are successfully acetylated. Also, castor oil methyl ester can be interesterified but with longer reaction time.
Sigma-Aldrich
Methyl acetate, anhydrous, 99.5%
Sigma-Aldrich
Methyl acetate, natural, 98%, FG
Supelco
Methyl acetate, analytical standard
Sigma-Aldrich
Methyl acetate, ≥98%, FG
Sigma-Aldrich
Methyl acetate, ReagentPlus®, 99%
Sigma-Aldrich
Methyl acetate, suitable for HPLC, ≥99.8%