Merck
  • Spectroscopic characterization studies of 1-methyl indole with benzene derivatives.
Spectroscopic characterization studies of 1-methyl indole with benzene derivatives.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2010-07-17)
S Bakkialakshmi, B Shanthi, M Shanthi
ABSTRACT
The NMR and fluorescence quenching of benzene, chlorobenzene and methoxybenzene in acetonitril and 2-propanol solvents have been carried out with a view to understand the quenching mechanisms. The quenching is found to be appreciable and shows positive derivation in the stern-volmer plots. UV absorbance is used to measure the excitation wavelength. Lifetime measurements are carried out and the quenching rate constant kq is also calculated. Molar extinction co-efficient, stoke's shift are also calculated. The NMR study is used to conform if there is any complex formation between 1-methylindole with the benzene derivatives.
Sigma-Aldrich
1-Methylindole, ≥97%
Sigma-Aldrich
Anisole, ≥99%, FCC, FG
Supelco
Anisole, analytical standard
Sigma-Aldrich
Anisole, anhydrous, 99.7%
Supelco
Anisole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anisole, ReagentPlus®, 99%