Merck
  • Palladium-catalyzed cross-coupling of cyclopropylmagnesium bromide with aryl bromides mediated by zinc halide additives.
Palladium-catalyzed cross-coupling of cyclopropylmagnesium bromide with aryl bromides mediated by zinc halide additives.
The Journal of organic chemistry (2010-09-08)
Chutian Shu, Kanwar Sidhu, Li Zhang, Xiao-Jun Wang, Dhileepkumar Krishnamurthy, Chris H Senanayake
ABSTRACT
The key Pd-catalyzed cross-coupling of aryl bromides or triflates and cyclopropylmagnesium bromide in the presence of substoichiometric amounts of zinc bromide produces cyclopropyl arenes in good to excellent yields. The cross-coupling of other alkyl, cycloalkyl, and aryl Grignard reagents with aryl bromides under the same conditions gives the corresponding substituted arenes in good yields.
Sigma-Aldrich
Zinc bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, puriss., anhydrous, ≥98%