Merck
  • Recombinant self-assembling peptides as biomaterials for tissue engineering.
Recombinant self-assembling peptides as biomaterials for tissue engineering.
Biomaterials (2010-10-12)
Stuart Kyle, Amalia Aggeli, Eileen Ingham, Michael J McPherson
ABSTRACT
Synthetic nanostructures based on self-assembling systems that aim to mimic natural extracellular matrix are now being used as substrates in tissue engineering applications. Peptides are excellent starting materials for the self-assembly process as they can be readily synthesised both chemically and biologically. P₁₁-4 is an 11 amino acid peptide that undergoes triggered self-assembly to form a self-supporting hydrogel. It exists as unimers of random coil conformations in water above pH 7.5 but at low pH adopts an antiparallel β-sheet conformation. It also self-assembles under physiological conditions in a concentration-dependent manner. Here we describe an unimer P₁₁-4 production system and the use of a simple site-directed mutagenesis approach to generate a series of other P₁₁-family peptide expression vectors. We have developed an efficient purification strategy for these peptide biomaterials using a simple procedure involving chemical cleavage with cyanogen bromide then repeated filtration, lyophilisation and wash steps. We report peptide-fusion protein yields of ca. 4.64 g/L and we believe the highest reported recovery of a recombinant self-assembling peptide at 203 mg/L of pure recombinant P₁₁-4. This peptide forms a self-supporting hydrogel under physiological conditions with essentially identical physico-chemical properties to the chemically synthesised peptide. Critically it also displays excellent cytocompatibility when tested with primary human dermal fibroblasts. This study demonstrates that high levels of a series of recombinant self-assembling peptides can be purified using a simple process for applications as scaffolds in tissue engineering.
Sigma-Aldrich
Cyanogen bromide, reagent grade, 97%
Sigma-Aldrich
Cyanogen bromide, 99.995% trace metals basis
Sigma-Aldrich
Cyanogen bromide, ≥98.5% (RT)