Merck
  • A nano-confined charged layer defies the principle of electrostatic interaction.
A nano-confined charged layer defies the principle of electrostatic interaction.
Chemical communications (Cambridge, England) (2011-05-20)
Prabhat K Singh, Manoj Kumbhakar, Haridas Pal, Sukhendu Nath
ABSTRACT
The reactivity between two charged molecules and the activity of charged biomolecules are mainly governed by the principle of electrostatic interaction, i.e., like charges repel and opposite charges attract. In the present study it is shown that the principle of electrostatic interaction is violated in the nano-confined biomimetic environment. Thus a positively charged molecule shows more preference to a positively charged surface compared to a negatively charged surface.
Sigma-Aldrich
Benzyldimethylhexadecylammonium chloride, cationic detergent