Merck
  • Monobenzone-induced depigmentation: from enzymatic blockade to autoimmunity.
Monobenzone-induced depigmentation: from enzymatic blockade to autoimmunity.
Pigment cell & melanoma research (2011-06-22)
Jasper G van den Boorn, Cornelis J Melief, Rosalie M Luiten
ABSTRACT
Autoimmune side-effects such as vitiligo regularly occur during melanoma immunotherapy. As vitiligo development is associated with a superior prognosis, the active induction of vitiligo in melanoma patients can be a useful tactic. The potent skin-depigmenting agent monobenzone can be used successfully for this purpose. However, until recently, the mechanism of action behind monobenzone-induced skin depigmentation was unclear. Lately, the mechanistic basis for the augmented immunogenicity of monobenzone-exposed pigmented cells has been unveiled, and their active role in the induction of autoimmune T-cell-mediated vitiligo has become apparent. Here, we provide an immunological framework in which we condense this knowledge to an integrated theory of the generation of monobenzone-induced vitiligo.
Sigma-Aldrich
4-(Benzyloxy)phenol, 98%
Pricing and availability is not currently available.