Merck
  • [Chlorination byproducts formation potentials of typical nitrogenous organic compounds in water].
[Chlorination byproducts formation potentials of typical nitrogenous organic compounds in water].
Huan jing ke xue= Huanjing kexue (2011-09-20)
Qian Xu, Bin Xu, Cao Qin, Sheng-Ji Xia, Nai-Yun Gao, Fu-Xiang Tian, Da-Peng Li
ABSTRACT
Twelve typical nitrogenous organic compounds including herbicides, pesticides, amino acids, industrial products etc in polluted raw water were selected to investigate formation of typical carbonaceous and nitrogenous DBPs during chlorination and chloramination. To indentify the formation mechanism of carbonaceous and nitrogenous disinfection byproducts from nitrogenous chemicals, chlorination and chloroamination of urea herbicides, triazine herbicides, amino acid, and other compounds were investigated. As a result, the potential precursors for different DBPs were defined as well. It has been identified that widely used urea herbicides could produce as many as 9 specific DBPs. The chlorotoluron shows highest reactivity and yields chloroform (CF), monochloroacetic acid (MCAA), dichloroacetic acid (DCAA), 1,1-dichloro-acetone (1,1-DCP), 1,1,1-trichloro-acetone (1,1,1-TCP), chloropicrin (NTCM), dichloro-acetonitrile (DCAN), dimethylnitrosamine (NDMA). The results indicated that aldicarb and dinoseb are important precursors of CF, DCAA, MCAA, NTCM as well. High concentrations of CF and DCAA were found during L-tryptophan chlorination. Furthermore, DBPs formation pathways and mechanisms were suggested during chlorination and chloramination of chlorotoluron, ametryn, dinoseb L-tryptophan.
Sigma-Aldrich
Sodium chloroacetate, 98%
Supelco
Chloroacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Chloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroacetic acid, 99%