Merck
  • Pi2 spacer pyrocarbon arthroplasty technique for thumb basal joint osteoarthritis.
Pi2 spacer pyrocarbon arthroplasty technique for thumb basal joint osteoarthritis.
Techniques in hand & upper extremity surgery (2011-11-23)
Philippe Bellemère, Ludovic Ardouin
ABSTRACT
Trapeziectomy associated with a pyrocarbon implant is a recent procedure for thumb basal joint osteoarthritis. The investigators report Pi pyrocarbon implant technique (Tornier Bioprofile, Grenoble, France) for primary thumb basal joint osteoarthritis. The key points of the procedure are preservation of the soft tissue environment during the trapeziectomy, partial trapezoidectomy to medialize the implant, and careful capsuloplasty and ligamentoplasty to stabilize the implant.
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)