Merck
  • Uliginosin B, a phloroglucinol derivative from Hypericum polyanthemum: a promising new molecular pattern for the development of antidepressant drugs.
Uliginosin B, a phloroglucinol derivative from Hypericum polyanthemum: a promising new molecular pattern for the development of antidepressant drugs.
Behavioural brain research (2011-12-14)
Ana C Stein, Alice F Viana, Liz G Müller, Jéssica M Nunes, Eveline D Stolz, Jean-Claude Do Rego, Jean Costentin, Gilsane L von Poser, Stela M K Rates
ABSTRACT
In this study we have demonstrated that cyclohexane extract of Hypericum polyanthemum (POL) and its main phloroglucinol derivative uliginosin B (ULI) present antidepressant-like activity in rodent forced swimming test (FST). The involvement of monoaminergic neurotransmission on the antidepressant-like activity of ULI was evaluated in vivo and in vitro. POL 90 mg/kg (p.o.) and ULI 10 mg/kg (p.o.) reduced the immobility time in the mice FST without altering locomotion activity in the open-field test. The combination of sub-effective doses of POL (45 mg/kg, p.o.) and ULI (5 mg/kg, p.o.) with sub-effective doses of imipramine (10 mg/kg, p.o.), bupropion (3 mg/kg, p.o.) and fluoxetine (15 mg/kg, p.o.) induced a significant reduction on immobility time in FST. The pretreatment with SCH 23390 (15 μg/kg, s.c., dopamine D1 receptor antagonist), sulpiride (50 mg/kg, i.p., dopamine D2 receptor antagonist), prazosin (1mg/kg, i.p., α1-adrenoceptor antagonist), yohimbine (1mg/kg, i.p., α2-adrenoceptor antagonist) and pCPA (100 mg/kg/day, i.p., p-chlorophenilalanine methyl ester, inhibitor of serotonin synthesis, for four consecutive days) before ULI administration (10 mg/kg, p.o.) significantly prevented the anti-immobility effect in FST. ULI was able to inhibit synaptosomal uptake of dopamine (IC50 = 90 ± 38 nM), serotonin (IC50 = 252 ± 13 nM) and noradrenaline (280 ± 48 nM), but it did not bind to any of the monoamine transporters. These data firstly demonstrated the antidepressant-like effect of POL and ULI, which depends on the activation of the monoaminergic neurotransmission in a different manner from the most antidepressants.
Sigma-Aldrich
4-Chloro-L-phenylalanine
Sigma-Aldrich
4-Chloro-DL-phenylalanine