Merck
  • A rapid and sensitive CE method with field-enhanced sample injection and in-capillary derivatization for selenomethionine metabolism catalyzed by flavin-containing monooxygenases.
A rapid and sensitive CE method with field-enhanced sample injection and in-capillary derivatization for selenomethionine metabolism catalyzed by flavin-containing monooxygenases.
Electrophoresis (2010-10-01)
Xin Hai, Thomas Nauwelaers, Roger Busson, Erwin Adams, Jos Hoogmartens, Ann Van Schepdael
ABSTRACT
A rapid and sensitive electrophoretically mediated microanalysis method with field-enhanced sample injection (FESI) for in-capillary derivatization was developed to determine selenomethionine (SeMet) and selenomethionine selenoxide (SeOMet). Phthalic anhydride (PA) was selected as the derivatization reagent due to the fast reaction at room temperature and the stability of derivatives. The in-capillary derivatization was accomplished by electrophoretically mixing PA and sample plugs. PA reagent was introduced hydrodynamically into the capillary, whereas the sample solution was injected electrokinetically, thus allowing a selective preconcentration of the analytes by FESI. For FESI, the optimum sample solvent was 2 mM borate solution. The borate buffer was suitable for both in-capillary derivatization and separation of the derivatives. The combination of electrophoretically mediated microanalysis with FESI for in-capillary derivatization was successfully achieved with about 800-fold concentration sensitivity enhancement compared to direct CE-UV detection in the same setup. The present method is miniaturized and fully automated, which ensures the on-line derivatization, stacking, separation and detection in 10 min. Finally, the developed method was successfully applied to measure enzyme activities by analyzing the reaction mixtures of SeMet with human flavin-containing monooxygenases (FMO). The results showed that both FMO1 and FMO3, but not FMO5 could catalyze the Se-oxygenation of SeMet.
Sigma-Aldrich
Phthalic anhydride, ReagentPlus®, 99%
Sigma-Aldrich
Phthalic anhydride, ACS reagent, ≥99%