Merck
  • A highly sensitive fluorogenic chemodosimeter for rapid visual detection of phosgene.
A highly sensitive fluorogenic chemodosimeter for rapid visual detection of phosgene.
Chemical communications (Cambridge, England) (2012-01-10)
Xuanjun Wu, Zhisheng Wu, Yuhui Yang, Shoufa Han
ABSTRACT
A highly sensitive chemodosimeter was identified from a panel of rhodamine derivatives for rapid and visual detection of phosgene with a detection limit of 50 nM triphosgene. Visual detection of gaseous phosgene with chemodosimeter absorbed paper strips was demonstrated.
Sigma-Aldrich
Phosgene solution, ~20% in toluene
Sigma-Aldrich
Phosgene solution, 15 wt. % in toluene