Merck
New thymine-based derivative of nitrogen mustards.
Nucleosides, nucleotides & nucleic acids (2012-02-24)
Benjamin Boëns, Karine Teste, Amel Hadj-Bouazza, Jihane Ismaili, Rachida Zerrouki
ABSTRACT
This work deals with the synthesis of a new nitrogen mustard derivative based on thymine. To introduce the bis(2-chloroethyl)amine group to position 4 of the pyrimidine base, many strategies were explored and the desired compound was finally obtained, thanks to a synthetic pathway in five steps.
Sigma-Aldrich
Diethanolamine, ACS reagent, ≥98.5%
Supelco
Diethanolamine, analytical standard
Sigma-Aldrich
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Diethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Diethanolamine, reagent grade, ≥98.0%
Sigma-Aldrich
Diethanolamine hydrochloride, 98%