Merck
  • Charge reorganization energy and small polaron binding energy of rubrene thin films by ultraviolet photoelectron spectroscopy.
Charge reorganization energy and small polaron binding energy of rubrene thin films by ultraviolet photoelectron spectroscopy.
Advanced materials (Deerfield Beach, Fla.) (2012-03-10)
Steffen Duhm, Qian Xin, Shunsuke Hosoumi, Hirohiko Fukagawa, Kazushi Sato, Nobuo Ueno, Satoshi Kera
ABSTRACT
The hole–phonon coupling of a rubrene monolayer on graphite is measured by means of angle resolved ultraviolet photoelectron spectroscopy. Thus, the charge reorganization energy λ and the small polaron binding energy is determined, which allows insight into the nature of charge transport in condensed rubrene.
Sigma-Aldrich
Rubrene, powder
Sigma-Aldrich
Rubrene, sublimed grade, 99.99% trace metals basis
Sigma-Aldrich
Rubrene, ≥98%