Merck
Impact of antiepileptic drugs on genesis of psychosis.
Epilepsy & behavior : E&B (2012-03-13)
Takahiro Noguchi, Naofumi Fukatsu, Hiroko Kato, Tomohiro Oshima, Kousuke Kanemoto
ABSTRACT
Opinions regarding the impact of antiepileptic drugs (AEDs) on the genesis of psychotic symptoms are varied. To re-examine this issue, the records of adult patients with partial epilepsy and newly added AEDs were retrospectively surveyed. The types of newly added AEDs and clinical characteristics were compared between 38 patients with active psychosis and 212 without psychotic history during a follow-up period of 3 to 6 months after initiation of AED administration. Using multivariate logistic regression analysis, the significance of possible predictive variables for development of psychosis was evaluated, which demonstrated that use of zonisamide (ZNS) and phenytoin (PHT), presence of complex partial seizures (CPS), and low intelligence level were significantly correlated with psychosis. We concluded that ZNS and PHT are possible risk factors for development of psychosis along with clinical variables, including the presence of CPS and low intelligence level.
Sigma-Aldrich
Zonisamide sodium salt, ≥98% (HPLC), powder
Supelco
Zonisamide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®