Merck
  • Organocatalytic asymmetric domino sulfa-Michael-aldol reactions of 2-mercaptobenzaldehyde with α,β-unsaturated N-acylpyrazoles for the construction of thiochromane.
Organocatalytic asymmetric domino sulfa-Michael-aldol reactions of 2-mercaptobenzaldehyde with α,β-unsaturated N-acylpyrazoles for the construction of thiochromane.
Chemical communications (Cambridge, England) (2012-05-05)
Xiu-Qin Dong, Xin Fang, Hai-Yan Tao, Xiang Zhou, Chun-Jiang Wang
ABSTRACT
An efficient protocol for the direct construction of bioactive thiochromanes was developed via a catalytic asymmetric cascade sulfa-Michael-aldol reaction of 2-mercaptobenzaldehyde with α,β-unsaturated N-acyl imides. The key to the present methodology is introducing a pyrazole moiety as H-bond acceptor, which allowed for better organization and activation and hence higher enantioselectivity.
Product Description
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Supelco
Benzaldehyde, analytical standard