Merck
  • Porphyrin dyads linked by a rotatable 3,3'-biphenyl scaffold: a new binding motif for small ditopic molecules.
Porphyrin dyads linked by a rotatable 3,3'-biphenyl scaffold: a new binding motif for small ditopic molecules.
Organic & biomolecular chemistry (2012-05-15)
Amy R Mulholland, Pall Thordarson, Emily J Mensforth, Steven J Langford
ABSTRACT
Synthetic access to a set of metallo- and free base bis-porphyrins has been provided by a stepwise approach involving sequential peptide and Suzuki couplings. Linking these porphyrins through a 3,3'-biphenyl bridge enables cooperative binding to ditopic ligands such as the bipyridyls. Association constants and binding stoichiometry has been determined by spectroscopic/spectrophotometric means and the differences in the binding affinities of a small series of diaza ligands is discussed in the context of structural fit and microscopic association constants.
Sigma-Aldrich
4,4′−Dipyridyl, 98%